Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC (-)-JNJ-A07 | 2135640-92-3 | 98.0% | 578.96 g/mol | C28H26ClF3N2O6 | 5 MG
SDP

Catalog No. 5000221010
Encompass_Preferred

(-)-JNJ-A07 is the (-) enantiomer of JNJ-A07, a potent and selective dengue virus (DENV) inhibitor that disrupts viral replication by targeting the NS3-NS4B interaction. It exhibits low-nanomolar antiviral activity (EC50 = 31 nM) and is supplied for research use. Chemical identifiers include CAS 2135640-92-3, formula C28H26ClF3N2O6, molecular weight 578.96 g/mol; typical purity is ~98%.

  • Potent DENV inhibitor (EC50 31 nM).
  • Chiral (-) enantiomer for stereospecific studies.
  • High chemical purity (~98%).
  • Available as solid and 10 mM solution in DMSO across multiple sizes.
  • Well-defined chemical identity: CAS 2135640-92-3; formula C28H26ClF3N2O6.
  • For research use only, not for human or veterinary use.

Catalog No. 50-002-21010 Supplier Medchemexpress LLC Supplier No. HY139602C5MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

(-)-JNJ-A07 is the (-) enantiomer of JNJ-A07, a potent and selective dengue virus (DENV) inhibitor that disrupts viral replication by targeting the NS3-NS4B interaction. It exhibits low-nanomolar antiviral activity (EC50 = 31 nM) and is supplied for research use. Chemical identifiers include CAS 2135640-92-3, formula C28H26ClF3N2O6, molecular weight 578.96 g/mol; typical purity is ~98%.

  • Potent DENV inhibitor (EC50 31 nM).
  • Chiral (-) enantiomer for stereospecific studies.
  • High chemical purity (~98%).
  • Available as solid and 10 mM solution in DMSO across multiple sizes.
  • Well-defined chemical identity: CAS 2135640-92-3; formula C28H26ClF3N2O6.
  • For research use only, not for human or veterinary use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.