Learn More
Medchemexpress LLC (+)-JNJ-A07 | 2135640-93-4 | 98.46% | 578.96 | 25 MG
SDP
Catalog No. 5000348153
Encompass_Preferred
(+)-JNJ-A07 is a highly potent, orally active pan-serotype dengue virus inhibitor that targets the NS3-NS4B interaction. It demonstrates nanomolar to picomolar activity against 21 clinical isolates and exhibits a favorable pharmacokinetic profile, leading to significant efficacy against dengue virus infection in mouse models.
  • Targets NS3-NS4B interaction
  • Potent pan-serotype dengue virus inhibitor
  • Effective against 21 clinical isolates
  • Favorable pharmacokinetic profile
  • High barrier to resistance
  • Inhibits viral replication complex formation
Catalog No. 50-003-48153 Supplier Medchemexpress LLC Supplier No. 25MGHY139602C
(+)-JNJ-A07 is a highly potent, orally active pan-serotype dengue virus inhibitor that targets the NS3-NS4B interaction. It demonstrates nanomolar to picomolar activity against 21 clinical isolates and exhibits a favorable pharmacokinetic profile, leading to significant efficacy against dengue virus infection in mouse models.
  • Targets NS3-NS4B interaction
  • Potent pan-serotype dengue virus inhibitor
  • Effective against 21 clinical isolates
  • Favorable pharmacokinetic profile
  • High barrier to resistance
  • Inhibits viral replication complex formation