Learn More
Selleck Chemical LLC JNJ-A07-E1422-1G
SDP
Catalog No. 503134940
Encompass_Preferred
JNJ-A07 is a highly potent inhibitor of Dengue virus (DENV) with an EC50 of 0 1 nM in the DENV-2 strain (16681) in Vero cells It exhibits antiviral activity by preventing the formation of the viral replication complex through blocking the interaction between two viral proteins NS3 and NS4B
Catalog No. 50-313-4940 Supplier Selleck Chemical LLC Supplier No. E14221G
JNJ-A07 is a highly potent inhibitor of Dengue virus (DENV) with an EC50 of 0 1 nM in the DENV-2 strain (16681) in Vero cells It exhibits antiviral activity by preventing the formation of the viral replication complex through blocking the interaction between two viral proteins NS3 and NS4B