Learn More
Selleck Chemical LLC JNJ-A07-E1422-25MG
SDP
Catalog No. 503135779
Encompass_Preferred
JNJ-A07 is a highly potent inhibitor of Dengue virus (DENV) with an EC50 of 0 1 nM in the DENV-2 strain (16681) in Vero cells It exhibits antiviral activity by preventing the formation of the viral replication complex through blocking the interaction between two viral proteins NS3 and NS4B
Catalog No. 50-313-5779 Supplier Selleck Chemical LLC Supplier No. E142225MG
JNJ-A07 is a highly potent inhibitor of Dengue virus (DENV) with an EC50 of 0 1 nM in the DENV-2 strain (16681) in Vero cells It exhibits antiviral activity by preventing the formation of the viral replication complex through blocking the interaction between two viral proteins NS3 and NS4B