Learn More
Selleck Chemical LLC JNJ0966
SDP
Catalog No. 5000002871
Encompass_Preferred
JNJ0966 is a highly selective compound that inhibits activation of MMP-9 zymogen and subsequent generation of catalytically active enzyme It has no effect on MMP-1 MMP-2 MMP-3 MMP-9 or MMP-14 catalytic activity and did not inhibit activation of the highly related MMP-2 zymogen
Catalog No. 50-000-02871 Supplier Selleck Chemical LLC Supplier No. S56962MG
JNJ0966 is a highly selective compound that inhibits activation of MMP-9 zymogen and subsequent generation of catalytically active enzyme It has no effect on MMP-1 MMP-2 MMP-3 MMP-9 or MMP-14 catalytic activity and did not inhibit activation of the highly related MMP-2 zymogen