Learn More
Medchemexpress LLC HY-103482 5mg Medchemexpress, JNJ0966 CAS:315705-75-0 Purity:>98%
SDP
Catalog No. 501968121
Encompass_Preferred
Medchemexpress, HY-103482 5mg JNJ0966 CAS:315705-75-0 JNJ0966 is a highly selective MMP-9 zymogen inhibitor with an IC50 of 440 nM. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-8121 Supplier Medchemexpress LLC Supplier No. HY1034825MG
Medchemexpress, HY-103482 5mg JNJ0966 CAS:315705-75-0 JNJ0966 is a highly selective MMP-9 zymogen inhibitor with an IC50 of 440 nM. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.