Learn More
Medchemexpress LLC JNJ16259685 | 409345-29-5 | 325.40 g/mol | C20H23NO3 | 1 ML
SDP
Supplier:  Medchemexpress LLC HY10040710MM 1ML
Encompass_Preferred
JNJ16259685 is a potent, selective antagonist of the metabotropic glutamate receptor 1 (mGlu1) that inhibits synaptic mGlu1 activation with nanomolar potency. It is supplied for research use in powder and solvent formats and is often provided as a concentrated solution in DMSO for biochemical and cellular assays.
  • Selective mGlu1 receptor antagonist with nanomolar potency.
  • Useful for studying mGlu1-mediated signaling in vitro and in vivo.
  • Available as powder or solution to suit experimental workflows.
  • Stable under recommended storage conditions for long-term use.
  • Characterized by a defined molecular formula and molecular weight for reproducibility.
  • Accompanied by product documentation such as datasheets and lot-specific quality information.
Catalog No. 50-002-14878