Learn More
Medchemexpress LLC HY-100407 5mg Medchemexpress, JNJ16259685 CAS:409345-29-5 Purity:>98%
SDP
Catalog No. 501931817
Encompass_Preferred
Medchemexpress, HY-100407 5mg JNJ16259685 CAS:409345-29-5 JNJ16259685 is a selective antagonist of mGlu1 receptor , and inhibits the synaptic activation of mGlu1 in a concentration-dependent manner with IC 50 of 19 nM. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-1817 Supplier Medchemexpress LLC Supplier No. HY1004075MG
Medchemexpress, HY-100407 5mg JNJ16259685 CAS:409345-29-5 JNJ16259685 is a selective antagonist of mGlu1 receptor , and inhibits the synaptic activation of mGlu1 in a concentration-dependent manner with IC 50 of 19 nM. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.