Learn More
Cayman Chemical JNJ47117096trIfluoroACTAt 10mg
SDP
Catalog No. 502945530
Encompass_Preferred
A MELK inhibitor (IC50 = 37 nM); selective for MELK over CAMKIIδ, MNK2, CAMKIIγ, and MLCK (IC50s = 810, 760, 1,000, and 1,000 nM, respectively) but does inhibit FLT3 (IC50 = 18 nM); inhibits MELK autophosphorylation (IC50 = 0.52 µM) and MELK proteasome-dependent degradation in MCF-7 cells; induces cellular senescence and the ATM-mediated DNA damage response in MCF-7 cells
Catalog No. 50-294-5530 Supplier Cayman Chemical Supplier No. 4160510 MG
A MELK inhibitor (IC50 = 37 nM); selective for MELK over CAMKIIδ, MNK2, CAMKIIγ, and MLCK (IC50s = 810, 760, 1,000, and 1,000 nM, respectively) but does inhibit FLT3 (IC50 = 18 nM); inhibits MELK autophosphorylation (IC50 = 0.52 µM) and MELK proteasome-dependent degradation in MCF-7 cells; induces cellular senescence and the ATM-mediated DNA damage response in MCF-7 cells