Learn More
TARGETMOL CHEMICALS INC JNK-IN-7 5mg
SDP
Catalog No. NC1624955
Encompass_Preferred
JNK-IN-7 is a selective JNK1/2/3 inhibitor (IC50: 1.54/1.99/0.75 nM). It can also inhibit phosphorylation of c-Jun, which is a substrate of JNK kinase.
Catalog No. NC1624955 Supplier TARGETMOL CHEMICALS INC Supplier No. T35985MG
JNK-IN-7 is a selective JNK1/2/3 inhibitor (IC50: 1.54/1.99/0.75 nM). It can also inhibit phosphorylation of c-Jun, which is a substrate of JNK kinase.