Learn More
Medchemexpress LLC HY-15617 5mg Medchemexpress, JNK-IN-7 CAS:1408064-71-0 Purity:>98%
SDP
Catalog No. 502081750
Encompass_Preferred
Medchemexpress, HY-15617 5mg JNK-IN-7 CAS:1408064-71-0 JNK-IN-7 is a potent JNK inhibitor with IC50 of 1.5, 2 and 0.7 nM for JNK1, JNK2 and JNK3, respectively. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-1750 Supplier Medchemexpress LLC Supplier No. HY156175MG
Medchemexpress, HY-15617 5mg JNK-IN-7 CAS:1408064-71-0 JNK-IN-7 is a potent JNK inhibitor with IC50 of 1.5, 2 and 0.7 nM for JNK1, JNK2 and JNK3, respectively. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.