Learn More
TARGETMOL CHEMICALS INC JNK-IN-7 5MG
SDP
Catalog No. 502580201
Encompass_Preferred
Also available in 1 mg 2 mg 10 mg 25 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. JNK-IN-7 (JNK inhibitor) is a selective JNK1/2/3 inhibitor (IC50 1.54/1.99/0.75 nM). It can also inhibit phosphorylation of c-Jun which is a substrate of JNK kinase. purity: 99%
Catalog No. 50-258-0201 Supplier TARGETMOL CHEMICALS INC Supplier No. T3598
Also available in 1 mg 2 mg 10 mg 25 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. JNK-IN-7 (JNK inhibitor) is a selective JNK1/2/3 inhibitor (IC50 1.54/1.99/0.75 nM). It can also inhibit phosphorylation of c-Jun which is a substrate of JNK kinase. purity: 99%