Learn More
Selleck Chemical LLC JNK-IN-8 S4901-1g
SDP
Catalog No. 503111041
Encompass_Preferred
JNK-IN-8 (JNK Inhibitor XVI) is the first irreversible JNK inhibitor for JNK1 JNK2 and JNK3 with IC50 of 4 7 nM 18 7 nM and 1 nM 10-fold selectivity against MNK2 Fms and no inhibition to c-Kit Met PDGFR in A375 cell line
Catalog No. 50-311-1041 Supplier Selleck Chemical LLC Supplier No. S49011G
JNK-IN-8 (JNK Inhibitor XVI) is the first irreversible JNK inhibitor for JNK1 JNK2 and JNK3 with IC50 of 4 7 nM 18 7 nM and 1 nM 10-fold selectivity against MNK2 Fms and no inhibition to c-Kit Met PDGFR in A375 cell line