Learn More
Apexbio Technology LLC JNK-IN-8 1410880-22-6 10mM (in 1mL DMSO)
SDP
Catalog No. 5000062876
Encompass_Preferred
JNK-IN-8 (CAS 1410880-22-6) is a potent and specific inhibitor of c-Jun N-terminal kinase (JNK) isoforms JNK1 JNK2 and JNK3 It functions by forming a covalent bond with a conserved cysteine residue in JNKs inducing conformational alteration in the kinase activation loop consequently impairing substrate binding Its IC50 values for JNK1 JNK2 and JNK3 are 4 67 nM 18 7 nM and 0 98 nM respectively In cellular assays JNK-IN-8 prevention of c-Jun phosphorylation demonstrates robust selectivity towards the JNK signaling pathway thus making it valuable as a research tool in studying stress-related cellular responses and apoptosis
Catalog No. 50-000-62876 Supplier Apexbio Technology LLC Supplier No. A35205.1
JNK-IN-8 (CAS 1410880-22-6) is a potent and specific inhibitor of c-Jun N-terminal kinase (JNK) isoforms JNK1 JNK2 and JNK3 It functions by forming a covalent bond with a conserved cysteine residue in JNKs inducing conformational alteration in the kinase activation loop consequently impairing substrate binding Its IC50 values for JNK1 JNK2 and JNK3 are 4 67 nM 18 7 nM and 0 98 nM respectively In cellular assays JNK-IN-8 prevention of c-Jun phosphorylation demonstrates robust selectivity towards the JNK signaling pathway thus making it valuable as a research tool in studying stress-related cellular responses and apoptosis