Learn More
Medchemexpress LLC HY-13319 2mg , JNK-IN-8 CAS:1410880-22-6 Purity:>98%
SDP
Catalog No. 502259697
Encompass_Preferred
HY-13319 2mg JNK-IN-8 CAS: 1410880-22-6 JNK-IN-8 (JNK Inhibitor XVI) is a potent JNK inhibitor with IC50s of 4.7 nM, 18.7 nM, and 1 nM for JNK1, JNK2, and JNK3, respectively. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
This item is not returnable. View return policy
Catalog No. 50-225-9697 Supplier Medchemexpress LLC Supplier No. HY133192MG
HY-13319 2mg JNK-IN-8 CAS: 1410880-22-6 JNK-IN-8 (JNK Inhibitor XVI) is a potent JNK inhibitor with IC50s of 4.7 nM, 18.7 nM, and 1 nM for JNK1, JNK2, and JNK3, respectively. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.