Learn More
Selleck Chemical LLC JNK-IN-8 5mg 1410880-22-6
SDP
Supplier:  Selleck Chemical LLC S49015MG
Encompass_Preferred
JNK-IN-8 is the first irreversible JNK inhibitor for JNK1, JNK2 and JNK3 with IC50 of 4.7 nM, 18.7 nM and 1 nM, >10-fold selectivity against MNK2, Fms and no inhibition to c-Kit, Met, PDGFR?in A375 cell line. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. NC0853526