Learn More
Medchemexpress LLC JNK Inhibitor VIII 10mM 1mL | 894804-07-0 | 10MM 1ML
SDP
Supplier:  Medchemexpress LLC HY10759810MM 1ML
Encompass_Preferred
JNK Inhibitor VIII (TCS JNK 6o) is a c-Jun N-terminal kinases (JNK-1 -2 and -3) inhibitor with Ki values of 2 nM 4 nM 52 nM respectively and has IC50 values of 45 nM and 160 nM for JNK-1 and -2 respectively[1
Catalog No. 50-002-40688