Learn More
Selleck Chemical LLC JNK Inhibitor VIII 2mg
SDP
Supplier:  Selleck Chemical LLC S77942MG
Encompass_Preferred
JNK Inhibitor VIII (TCS JNK 6o) is an inhibitor ofc-Jun N-terminal kinases with IC50 of 45 nM and 160 nM for JNK-1 and JNK-2, respectively. JNK Inhibitor VIII (TCS JNK 6o) inihibits JNK-1, JNK-2, and JNK-3 with Ki of 2 nM, 4 nM and 52 nM, respectively. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-217-3308