Learn More
Medchemexpress LLC HY-107598 5mg Medchemexpress, JNK Inhibitor VIII CAS:894804-07-0 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1075985MG
Encompass_Preferred
Medchemexpress, HY-107598 5mg JNK Inhibitor VIII CAS:894804-07-0 JNK Inhibitor VIII (TCS JNK 6o) is a c-Jun N-terminal kinases (JNK-1, -2, and -3) inhibitor with Ki values of 2 nM, 4 nM, 52 nM, respectively, and has IC50 values of 45 nM and 160 nM for JNK-1 and -2, respectively. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-8435