Learn More
Selleck Chemical LLC JNK Inhibitor VIII
SDP
Catalog No. 5000004619
Encompass_Preferred
JNK Inhibitor VIII (TCS JNK 6o) is an inhibitor of c-Jun N-terminal kinases with IC50 of 45 nM and 160 nM for JNK-1 and JNK-2 respectively JNK Inhibitor VIII (TCS JNK 6o) inihibits JNK-1 JNK-2 and JNK-3 with Ki of 2 nM 4 nM and 52 nM respectively
Catalog No. 50-000-04619 Supplier Selleck Chemical LLC Supplier No. S7794100MG
Add to Cart
Add to Cart
JNK Inhibitor VIII (TCS JNK 6o) is an inhibitor of c-Jun N-terminal kinases with IC50 of 45 nM and 160 nM for JNK-1 and JNK-2 respectively JNK Inhibitor VIII (TCS JNK 6o) inihibits JNK-1 JNK-2 and JNK-3 with Ki of 2 nM 4 nM and 52 nM respectively