Learn More
Cayman Chemical JNK InhIbItr IX 10mg
SDP
Catalog No. 502968699
Encompass_Preferred
A thienylnaphthamide compound that selectively targets the ATP binding site of JNK2 and JNK3 (pIC50s = 6.5 and 6.7, respectively)
Catalog No. 50-296-8699 Supplier Cayman Chemical Supplier No. 1562410 MG
Add to Cart
Add to Cart
A thienylnaphthamide compound that selectively targets the ATP binding site of JNK2 and JNK3 (pIC50s = 6.5 and 6.7, respectively)