Learn More
Cayman Chemical JNK InhIbItr V 1mg
SDP
Catalog No. 502965836
Encompass_Preferred
An ATP-competitive inhibitor of JNK1, JNK2, and JNK3 (IC50s = 150, 220, and 70 nM, respectively); effective in vivo in gerbils, mice, and rats via oral, intravenous, or intraperitoneal administration
Catalog No. 50-296-5836 Supplier Cayman Chemical Supplier No. 175421 MG
An ATP-competitive inhibitor of JNK1, JNK2, and JNK3 (IC50s = 150, 220, and 70 nM, respectively); effective in vivo in gerbils, mice, and rats via oral, intravenous, or intraperitoneal administration