Learn More
Cayman Chemical JNK Inhibitor V, Purity: ≥98% (sum of isomers), CAS: 345987-15-7, 25 mg
SDP
Catalog No. 502966662
Encompass_Preferred
An ATP-competitive inhibitor of JNK1, JNK2, and JNK3 (IC50s = 150, 220, and 70 nM, respectively); effective in vivo in gerbils, mice, and rats via oral, intravenous, or intraperitoneal administration
Catalog No. 50-296-6662 Supplier Cayman Chemical Supplier No. 1754225 MG
An ATP-competitive inhibitor of JNK1, JNK2, and JNK3 (IC50s = 150, 220, and 70 nM, respectively); effective in vivo in gerbils, mice, and rats via oral, intravenous, or intraperitoneal administration