Learn More
Cayman Chemical JNK InhIbItr VII 25mg
SDP
Catalog No. 502938331
Encompass_Preferred
A non-selective JNK inhibitor (IC50s = 1.54, 1.99, and 0.75 nM for JNK1, -2, and -3, respectively); inhibits phosphorylation of c-Jun in HeLa and A375 cells (EC50s = 130 and 244 nM, respectively)
Catalog No. 50-293-8331 Supplier Cayman Chemical Supplier No. 2883125 MG
Add to Cart
Add to Cart
A non-selective JNK inhibitor (IC50s = 1.54, 1.99, and 0.75 nM for JNK1, -2, and -3, respectively); inhibits phosphorylation of c-Jun in HeLa and A375 cells (EC50s = 130 and 244 nM, respectively)