Learn More
Cayman Chemical JNK InhIbItr XVI 1mg
SDP
Catalog No. 502979820
Encompass_Preferred
A selective, irreversible JNK inhibitor (IC50s = 4.67, 18.7, and 0.98 nM for JNK1, 2, and 3, respectively); prevents phosphorylation of c-Jun in A375 and HeLa cells (EC50s = 338 and 486 nM, respectively)
Catalog No. 50-297-9820 Supplier Cayman Chemical Supplier No. 180961 MG
A selective, irreversible JNK inhibitor (IC50s = 4.67, 18.7, and 0.98 nM for JNK1, 2, and 3, respectively); prevents phosphorylation of c-Jun in A375 and HeLa cells (EC50s = 338 and 486 nM, respectively)