Learn More
Enzo Life Sciences Jnk1/2 (pThr183/Tyr185) ELISA kit (1x96wells)
SDP
Supplier:  Enzo Life Sciences ADI900106
Encompass_Preferred
The [pThr183/Tyr185]Jnk1/2 EIA kit is a colorimetric immunometric enzyme immunoassay kit with results in 3 hours. Absorbance is read at 450 nm. Save time, money, and precious sample with fully quantitative results and increased sensitivity compared to Western blot analysis. Alternative Name: c-Jun N-terminal kinase 1/2. SAP kinase 1a/c. MAP kinase 8/9. MAPK8/9. Sensitivity: 75.8 pg/ml (range 125 - 4,000 pg/ml). Assay Time: 3 hours. Applications: ELISA, Colorimetric detection. Species Reactivity: Human, Mouse, Rat. Contents: Microtiter plate, Conjugate, Antibody, Assay buffer 4 concentrate, Wash buffer concentrate, Standard, TMB Substrate, RIPA Cell lysis buffer 2, Stop solution 2
Catalog No. 50-658-145