Learn More
Enzo Life Sciences JNK1/2 (pThr183/pTyr185) polyclonal antibody (20 µl)
SDP
Supplier:  Enzo Life Sciences BMLSA2720020
Encompass_Preferred
Immunogen: Synthetic peptide corresponding to a portion of human JNK1&2 phosphorylated at Thr183 and Tyr185. This region is conserved among many species, including mouse, rat, chicken, nematode and fly. Species reactivity: Human, Rat. Specificity: Recognizes JNK 1&2 dually phosphorylated at Thr183 and Tyr185. Applications: IHC, WB. Formulation: Liquid. In PBS containing BSA and sodium azide.
Catalog No. 50-201-1787