Learn More
Carnabio Usa Inc JNK1(MAPK8)/5UG/CATALYTIC
Supplier:  Carnabio Usa Inc 041635
Encompass
JNK1(MAPK8), Catalytic domain, Wild type, Amino Acid 2-364, NP_620634.1, Expressed in E. coli; 5microg
Catalog No. 50-754-0474