Learn More
Sino Biological JNK2 / MAPK9 Antibody, Mouse MAb 100uL
Supplier:  Sino Biological 10745MM01100
Encompass_Preferred
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Human MAPK9 (rh MAPK9; Catalog#10745-H08B; NP_002743.3; Met 1-Arg 424). The IgG fraction of the cell culture supernatant was purified by Protein A affinity chromatography.
Catalog No. 50-224-8556