Learn More
Sino Biological JNK2 / MAPK9 Antibody, Rabbit MAb 100uL
Supplier:  Sino Biological 10745R011100
Encompass_Preferred
This antibody was obtained from a rabbit immunized with purified, recombinant Human JNK2 / MAPK9 (rh JNK2 / MAPK9; Catalog#10745-H08B; NP_002743.3; Met 1-Arg 424).
Catalog No. 50-224-7850