Learn More
Sino Biological JNK2 / MAPK9 Antibody, Rabbit PAb, Antigen Affinity Purified 100uL
Supplier:  Sino Biological 10745T20100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Human JNK2 / MAPK9 (rh JNK2 / MAPK9; Catalog#10745-H08B; NP_002743.3; Met1-Arg424). JNK2 / MAPK9 specific IgG was purified by Human JNK2 / MAPK9 affinity chromatography.
Catalog No. 50-224-8558