Learn More
Eurofins Discoverx JNK3/SAPK1b, unactive, 50 µg. (14-523)
Supplier:  Eurofins Discoverx 14523
Encompass
N-terminal 6xHis-tagged recombinant, human JNK3/SAPK1b. For detailed product information, visit www.discoverx.com/proteins.
Catalog No. 50-199-8915