Learn More
Chemglass Life Sciences Clamp, Keck, Yellow, Standard Taper, Fits Joint Sizes 14/20 & 14/35
SDP
Catalog No. CG14503
Encompass_Preferred
Clamp, Keck, Yellow, Standard Taper, Fits Joint Sizes 14/20 & 14/35. Made of Delrin, the clamp is non-corrosive and will not scratch glass. Can be used to 140 C. Color coded for easy identification. SOLD IN PACKAGES OF 10.
Catalog No. CG14503 Supplier Chemglass Life Sciences Supplier No. CG14503
Clamp, Keck, Yellow, Standard Taper, Fits Joint Sizes 14/20 & 14/35. Made of Delrin, the clamp is non-corrosive and will not scratch glass. Can be used to 140 C. Color coded for easy identification. SOLD IN PACKAGES OF 10.