Learn More
Cayman Chemical JnthItrem A 2.5mg
SDP
Catalog No. 502991307
Encompass_Preferred
A mycotoxin; induces tremors in mice and decreases the time to fall from the rotarod at 4 mg/kg; reduces W. cervinata larvae weight gain and food consumption at 20 UG/g
Catalog No. 50-299-1307 Supplier Cayman Chemical Supplier No. 301662.5 MG
A mycotoxin; induces tremors in mice and decreases the time to fall from the rotarod at 4 mg/kg; reduces W. cervinata larvae weight gain and food consumption at 20 UG/g