Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Brady™ Job Site Binder Only

Catalog No. 19103209 Shop All Brady Corporation Products
Change view
Click to view available options
Width (Metric):
311.15 mm
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Width (Metric)
19-103-209 311.15 mm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 19-103-209 Supplier Brady™ Supplier No. 2022

Carry your Material Safety Data Sheets with you using our convenient Job Site Binder with handles

Specifications

Format Binder
Includes Binder
Diameter (English) Ring 2 in.
Width (Metric) 311.15 mm
Height (English) 11 in.
Quantity Each
Depth (English) 2 in.
Material Polyethylene
Color Yellow on Red
Languages English
For Use With (Application) Hazard Communication
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.