Learn More
MSC M.A. Ford Drill Bit, 1/2 in, Altima, 140 Point
Catalog No. NC3637793
Encompass
The M.A. Ford HPDSR series Jobber Length Drill Bit is designed for high-performance drilling in a wide range of materials. It features a self-centering point and a durable Altima coating, making it suitable for demanding job shop and batch production work.
  • Self-centering point eliminates spot drilling
  • Corner chamfer angle of 45° minimizes burrs and aids material breakout
  • H7 OD tolerance for precision holes
  • Altima coating for high toughness and low friction
  • Solid carbide construction
  • Five times diameter (5xD) drilling length
  • Point angle is 140°
  • H6 Shank OD tolerance for shrink fit applications
  • Two flutes
Catalog No. NC3637793 Supplier MSC Supplier No. 12441325
The M.A. Ford HPDSR series Jobber Length Drill Bit is designed for high-performance drilling in a wide range of materials. It features a self-centering point and a durable Altima coating, making it suitable for demanding job shop and batch production work.
  • Self-centering point eliminates spot drilling
  • Corner chamfer angle of 45° minimizes burrs and aids material breakout
  • H7 OD tolerance for precision holes
  • Altima coating for high toughness and low friction
  • Solid carbide construction
  • Five times diameter (5xD) drilling length
  • Point angle is 140°
  • H6 Shank OD tolerance for shrink fit applications
  • Two flutes