Learn More
AVANTOR MASTERFLEX (VWR LLC) John Guest PPSV040808W Speedfit Ball Valve, straight, 1/4"" OD speedfit x 1/4"" OD speedfit
Catalog No. NC9418173
Encompass
Food-grade polypropylene housing is specifically made for use with potable water. Quickly disconnect and reconnect your water lines with these Speedfit valves.
Catalog No. NC9418173 Supplier AVANTOR MASTERFLEX (VWR LLC) Supplier No. MFLX0739104
Food-grade polypropylene housing is specifically made for use with potable water. Quickly disconnect and reconnect your water lines with these Speedfit valves.