Learn More
Roboz Surgical JOHNS HOPKINS CLAMP 1.5IN STR
SDP
Supplier:  Roboz Surgical RS7440
Encompass
Johns Hopkins Bulldog Clamp; Straight; 700 Grams Pressure; 1.5 in. Length The Bulldog clamp is a useful instrument for stopping blood flow to a particular organ of interest. One of the most common applications where a bulldog clamp can be used is
Catalog No. NC9767190