Learn More
MSC Johnson Level T-Square, 48 in, Aluminum, 0.19 in Blade
Catalog No. 5000529334
Encompass
The Johnson Level T-Square is a durable 48 in aluminum drywall T-square featuring an extra-thick blade and a high-strength riveting process for accurate measuring and marking. It is designed to match the height of standard drywall or plywood panels, offering large, easy-to-read graduations and a clear, protective anodized finish.
  • Heavy-duty, heat-treated aluminum construction with a robust 3/16 inch blade.
  • High-strength riveting process for accurate measuring and marking.
  • Easy-to-read 1/8 inch thermal-bonded graduations.
  • Blade graduations readable in both directions with 16 inch center markings.
  • Anodized finish protects against rust and corrosion.
  • Designed to match the height of typical drywall or plywood panels.
  • Sturdy aluminum blade functions as a straight edge, preventing utility knife slippage.
  • Lightweight construction for easy handling.
Catalog No. 50-005-29334 Supplier MSC Supplier No. 06290647
The Johnson Level T-Square is a durable 48 in aluminum drywall T-square featuring an extra-thick blade and a high-strength riveting process for accurate measuring and marking. It is designed to match the height of standard drywall or plywood panels, offering large, easy-to-read graduations and a clear, protective anodized finish.
  • Heavy-duty, heat-treated aluminum construction with a robust 3/16 inch blade.
  • High-strength riveting process for accurate measuring and marking.
  • Easy-to-read 1/8 inch thermal-bonded graduations.
  • Blade graduations readable in both directions with 16 inch center markings.
  • Anodized finish protects against rust and corrosion.
  • Designed to match the height of typical drywall or plywood panels.
  • Sturdy aluminum blade functions as a straight edge, preventing utility knife slippage.
  • Lightweight construction for easy handling.