Learn More
MSC Johnson Level & Tool Combination Square Set, 2 Piece, 12 in, Inch/Metric
Catalog No. 5000535556
Encompass
This combination square set is a two-piece, 12 in tool. It features inch/metric graduation, a durable stainless steel blade, and a robust cast zinc head.
  • Two pieces
  • 12 in blade
  • Inch/metric graduation
  • Graduation in 1/8 in, 1/16 in, and 1/32 in increments
  • Stainless steel blade material
  • Cast zinc head material
Catalog No. 50-005-35556 Supplier MSC Supplier No. 50312453
This combination square set is a two-piece, 12 in tool. It features inch/metric graduation, a durable stainless steel blade, and a robust cast zinc head.
  • Two pieces
  • 12 in blade
  • Inch/metric graduation
  • Graduation in 1/8 in, 1/16 in, and 1/32 in increments
  • Stainless steel blade material
  • Cast zinc head material