Learn More
MSC Johnson Level & Tool Digital Angle Locator, 0-180°, 0.1° Resolution, Magnetic Base
Catalog No. 5000531004
Encompass
This digital angle locator is designed for easily and accurately checking and transferring angle measurements. It displays angles in degrees with a working range of 0-180 degrees and an accuracy of ± 0.1°. The magnetic base makes it useful for various applications.
  • Displays angles in degrees
  • Working range of angle measurement 0-180 degrees
  • Magnetic base for hands-free use on ferrous surfaces
  • Accuracy of ± 0.1°
  • Resolution of 0.1°
  • Includes a soft-sided pouch for protection
  • Ideal for remodeling, cabinet installation, and measuring roof angles
Catalog No. 50-005-31004 Supplier MSC Supplier No. 69820389
This digital angle locator is designed for easily and accurately checking and transferring angle measurements. It displays angles in degrees with a working range of 0-180 degrees and an accuracy of ± 0.1°. The magnetic base makes it useful for various applications.
  • Displays angles in degrees
  • Working range of angle measurement 0-180 degrees
  • Magnetic base for hands-free use on ferrous surfaces
  • Accuracy of ± 0.1°
  • Resolution of 0.1°
  • Includes a soft-sided pouch for protection
  • Ideal for remodeling, cabinet installation, and measuring roof angles