Learn More
MSC Johnson Level & Tool Line Laser Level, 2 Beams, Class II, Red Beam
Catalog No. 5000527178
Encompass
The Johnson Level & Tool Line Laser Level is a versatile tool designed for various leveling applications. It features 2 beams, is classified as Class II, and emits a red laser beam, offering precise measurements with an accuracy of 1/4 inch at 20 feet. Powered by 2 AA alkaline batteries, it is suitable for tasks such as floor installation.
  • Line laser type
  • Class II laser
  • 1/4 inch at 20 feet accuracy
  • Battery powered
  • Includes 2 AA alkaline batteries
  • Red beam color
  • Suitable for floor installation
  • Dual leveling vial
Catalog No. 50-005-27178 Supplier MSC Supplier No. 44538106
The Johnson Level & Tool Line Laser Level is a versatile tool designed for various leveling applications. It features 2 beams, is classified as Class II, and emits a red laser beam, offering precise measurements with an accuracy of 1/4 inch at 20 feet. Powered by 2 AA alkaline batteries, it is suitable for tasks such as floor installation.
  • Line laser type
  • Class II laser
  • 1/4 inch at 20 feet accuracy
  • Battery powered
  • Includes 2 AA alkaline batteries
  • Red beam color
  • Suitable for floor installation
  • Dual leveling vial