Learn More
MSC Johnson Level & Tool Steel Rule, 96 in OAL, English Graduation, Anodized Aluminum
Catalog No. 5000535065
Encompass
This aluminum straight edge is designed for durability and accuracy, featuring a 96-inch overall length. Its surface is enhanced with screened, thermal-bonded English graduations in 1/8 inch and 1/16 inch for clear and easy readability. A clear anodized finish protects the rule, and a convenient hanging hole is included for storage. The rigid, 2-inch wide design ensures stability during use.
  • Heavy-duty construction ensures lasting performance.
  • 96-inch overall length provides extensive measurement capability.
  • Durable clear anodized finish protects against wear.
  • Screened, thermal-bonded English graduations for precise measurements.
  • Bold and easy-to-read 1/8 inch and 1/16 inch markings.
  • Integrated hanging hole for convenient storage.
  • Rigid, 2-inch wide design ensures stability.
  • Not NIST certified.
Catalog No. 50-005-35065 Supplier MSC Supplier No. 56309552
This aluminum straight edge is designed for durability and accuracy, featuring a 96-inch overall length. Its surface is enhanced with screened, thermal-bonded English graduations in 1/8 inch and 1/16 inch for clear and easy readability. A clear anodized finish protects the rule, and a convenient hanging hole is included for storage. The rigid, 2-inch wide design ensures stability during use.
  • Heavy-duty construction ensures lasting performance.
  • 96-inch overall length provides extensive measurement capability.
  • Durable clear anodized finish protects against wear.
  • Screened, thermal-bonded English graduations for precise measurements.
  • Bold and easy-to-read 1/8 inch and 1/16 inch markings.
  • Integrated hanging hole for convenient storage.
  • Rigid, 2-inch wide design ensures stability.
  • Not NIST certified.