Learn More
MSC 80/20 Inc. Joining Strip, 2 Hole, Aluminum, 2 in., 1 ea.
Catalog No. 5000547772
Encompass
This 2-hole straight flat plate serves as an external fastening method for 10 series profiles. Constructed from durable 6063-T6 aluminum with an anodized finish, this component is designed for use with bolt kit 3321. It provides a secure connection that allows for infinite positioning along the profile T-slot and can be easily repositioned or reused as needed.
  • Features 2-degree drop-lock functionality for a strong connection when assembly bolts are tightened
  • Manufactured from 6063-T6 aluminum
  • Allows for infinite positioning along the profile T-slot
  • Easily repositioned and reused
  • ESD safe design
  • Measures 2 in. length by 0.875 in. width with a 0.188 in. thickness
Catalog No. 50-005-47772 Supplier MSC Supplier No. 76824465
This 2-hole straight flat plate serves as an external fastening method for 10 series profiles. Constructed from durable 6063-T6 aluminum with an anodized finish, this component is designed for use with bolt kit 3321. It provides a secure connection that allows for infinite positioning along the profile T-slot and can be easily repositioned or reused as needed.
  • Features 2-degree drop-lock functionality for a strong connection when assembly bolts are tightened
  • Manufactured from 6063-T6 aluminum
  • Allows for infinite positioning along the profile T-slot
  • Easily repositioned and reused
  • ESD safe design
  • Measures 2 in. length by 0.875 in. width with a 0.188 in. thickness