Learn More
Chemglass Life Sciences 10/30 Outer Joint, Full Length, on 14mm OD x 1.2mm W tubing
SDP
Catalog No. 501530985
Encompass_Preferred
10/30 Outer Joint, Full Length, on 14mm OD x 1.2mm W tubing. Standard taper outer joint tooled on tubing approximately the same diameter as the top reinforcing ring, affording no hold-up below joint.
Catalog No. 50-153-0985 Supplier Chemglass Life Sciences Supplier No. CG10102
10/30 Outer Joint, Full Length, on 14mm OD x 1.2mm W tubing. Standard taper outer joint tooled on tubing approximately the same diameter as the top reinforcing ring, affording no hold-up below joint.