Learn More
Chemglass Life Sciences O-Ring Joint, 12mm ID, 16mm Tube OD , O-Ring Size: 116, Clamp Size: 28
SDP
Catalog No. 501531068
Encompass_Preferred
O-Ring Joint, 12mm ID, 16mm Tube OD , O-Ring Size: 116, Clamp Size: 28. Joint has an o-ring groove to accept the listed o-ring size. Provides for a grease-free, vacuum seal when used with CG-150 clamps. Each joint is supplied with a Viton o-ring of the listed size.
Catalog No. 50-153-1068 Supplier Chemglass Life Sciences Supplier No. CG12420
O-Ring Joint, 12mm ID, 16mm Tube OD , O-Ring Size: 116, Clamp Size: 28. Joint has an o-ring groove to accept the listed o-ring size. Provides for a grease-free, vacuum seal when used with CG-150 clamps. Each joint is supplied with a Viton o-ring of the listed size.