Learn More
Chemglass Life Sciences 14/20 Inner Joint, Cut Drip, on 12.7mm OD x 1.6mm
SDP
Catalog No. 501531029
Encompass_Preferred
14/20 Inner Joint, Cut Drip, on 12.7mm OD x 1.6mm. Standard taper inner joint having a drip tip. Joints are available uncut or with the drip tip being cut at a 45 angle. Tooled on the same tubing sizes as the CG-100 inner joints.
Catalog No. 50-153-1029 Supplier Chemglass Life Sciences Supplier No. CG11702
14/20 Inner Joint, Cut Drip, on 12.7mm OD x 1.6mm. Standard taper inner joint having a drip tip. Joints are available uncut or with the drip tip being cut at a 45 angle. Tooled on the same tubing sizes as the CG-100 inner joints.