Learn More
Chemglass Life Sciences 24/40 Inner Joint, Cut Drip, on 22mm OD x 2.5mm W tubing
SDP
Catalog No. 501531031
Encompass_Preferred
24/40 Inner Joint, Cut Drip, on 22mm OD x 2.5mm W tubing. Full or medium length standard taper inner joint having a drip tip. Joints are available uncut or with the drip tip being cut at a 45 angle. Tooled on the same tubing sizes as the CG-100 inner joints.
Catalog No. 50-153-1031 Supplier Chemglass Life Sciences Supplier No. CG11714
24/40 Inner Joint, Cut Drip, on 22mm OD x 2.5mm W tubing. Full or medium length standard taper inner joint having a drip tip. Joints are available uncut or with the drip tip being cut at a 45 angle. Tooled on the same tubing sizes as the CG-100 inner joints.