Learn More
Chemglass Life Sciences 24/40 Outer Joint with Hooks, on 28mm OD x 2mm W tubing
SDP
Catalog No. 501531002
Encompass_Preferred
24/40 Outer Joint with Hooks, on 28mm OD x 2mm W tubing. Outer joint with two hooks 180 apart just below the grind.
Catalog No. 50-153-1002 Supplier Chemglass Life Sciences Supplier No. CG10808
24/40 Outer Joint with Hooks, on 28mm OD x 2mm W tubing. Outer joint with two hooks 180 apart just below the grind.