Learn More
Chemglass Life Sciences 35/25 O-Ring Ball Joint, Clamp Size: 35, O-Ring Size: 120, Tubing OD: 28mm
SDP
Catalog No. 50873588
Encompass_Preferred
35/25 O-Ring Ball Joint, Clamp Size: 35, O-Ring Size: 120,Tubing OD: 28mm. Ball joint with a Viton o-ring fitted into a groove tooled at the midpoint of the grind. Provides for a flexible, vacuum tight seal when used with CG-121-P polished socket joints. Supplied complete with one Viton o-ring of the listed size.
Catalog No. 50-873-588 Supplier Chemglass Life Sciences Supplier No. CG12209
35/25 O-Ring Ball Joint, Clamp Size: 35, O-Ring Size: 120,Tubing OD: 28mm. Ball joint with a Viton o-ring fitted into a groove tooled at the midpoint of the grind. Provides for a flexible, vacuum tight seal when used with CG-121-P polished socket joints. Supplied complete with one Viton o-ring of the listed size.